missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T7 RNA Polymerase (20 U/μL)

Catalog No. FEREP0112
Change view
Click to view available options
Concentration:
20U/µL
Quantity:
5 x 5,000 units
5,000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Quantity
FEREP0112 20U/µL 5 x 5,000 units
FEREP0111 20U/µL 5,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEREP0112 Supplier Thermo Scientific™ Supplier No. EP0112

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ T7 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.

Thermo Scientific™ Bacteriophage T7 RNA polymerase is DNA-dependent RNA polymerase with strict specificity its respective double-stranded promoter.

It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)

Recommended for:

Synthesis of unlabeled and labeled RNA that can be used:for hybridization, in vitro RNA translation; as aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing.

Note:

Consensus promoter sequences: TAATACGACTCACTATA G GGAGA. The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively.

Specifications

Polymerase T7 RNA Polymerase
Quantity 5 x 5,000 units
Concentration 20U/µL
What is the advantage of using TranscriptAid T7 High Yield Transcription Kit over Thermo Scientific T7 RNA Polymerase with a standard transcription buffer?

TranscriptAid T7 High Yield Transcription Kit is designed to produce both long and short transcripts for applications that require large yields of RNA (milligram amounts). The kit can achieve 10 times greater amount than from conventional in vitro transcription reactions with T7 RNA polymerase due to its unique proprietary enzyme and buffer formulations. Please note that TranscriptAid T7 High Yield Transcription Kit is not recommended for generation of radioactively labelled RNA. Due to large quantities of RNA synthesized with the kit, generation of high specific activity radiolabeled probes would require prohibitively large amounts of radiolabeled nucleotide.

For Research Use Only. Not for use in diagnostic procedures.