missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RevertAid Reverse Transcriptase (200 U/μL)

Thermo Scientific RevertAid Reverse Transcriptase (RT) is a recombinant M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties.

Supplier:  Thermo Scientific™ EP0442

 View more versions of this product

Catalog No. FEREP0442


Only null left

Description

Description

Thermo Scientific RevertAid Reverse Transcriptase (RT) is a recombinant M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids that is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.

Features of RevertAid Reverse Transcriptase include:
• Efficient synthesis of full-length first strand cDNA up to 13 kb
• Optimum activity at 42°C
• Active up to 50°C
• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, and fluorescein-labeled nucleotides)

Applications
• First strand cDNA synthesis for RT-PCR and real-time RT-PCR
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• DNA labeling
• Analysis of RNA by primer extension

Source
RevertAid Reverse Transcriptase is sourced from E.coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus reverse transcriptase.

Specifications

Specifications

200 U/μL
Stand-alone Enzyme
Reverse Transcription
5 x 10,000 Units
Yes
First-Strand cDNA
Reverse Transcription
RNA

• RevertAid Reverse Transcriptase (200 U/μL)
• 5X Reaction Buffer

Store at –20°C.

60 min.
42°C
RevertAid
Dry Ice
Separate components
Up to 13 kb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.