missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RevertAid First Strand cDNA Synthesis Kit

Catalog No. FERK1621
Change view
Click to view available options
No. of Reactions:
100 Reactions
20 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
FERK1621 20 Reactions
FERK1622 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK1621 Supplier Thermo Scientific™ Supplier No. K1621

Thermo Scientific RevertAid First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates.

Thermo Scientific RevertAid First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates. The kit uses RevertAid Reverse Transcriptase (RT), a recombinant M-MuLV RT which maintains activity at 42–50°C and is suitable for synthesis of cDNA up to 13 kb. RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA templates from degradation at temperatures up to 55°C.

The RevertAid First Strand cDNA Synthesis Kit is supplied with both oligo(dT)18 and random hexamer primers. The oligo(dT)18 primer anneals selectively to the poly(A) tail of mRNA. Random hexamer primers do not require the presence of the poly(A) tail; they can therefore be used for transcription of the 5'-end regions of mRNA or cDNA synthesis of RNA species lacking a poly(A) tail (e.g., microRNAs). Gene-specific primers may also be used with the kit.

Radioactively- and non-radioactively-labeled nucleotides can be incorporated into first strand cDNA for use as a probe in hybridization experiments, including microarrays.

Features of the RevertAid First Strand cDNA Synthesis Kit include:
• Full-length first strand cDNA up to 13 kb
• Optimum reaction temperature 42°C
• Complete kit with all components necessary for the RT reaction already included

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

Specifications

Content And Storage

• RevertAid Reverse Transcriptase
• RiboLock RNase Inhibitor
• 5X Reaction Buffer
• dNTP Mix
• Oligo(dT)18 Primer
• Random Hexamer Primer
• Control GAPDH RNA
• Forward GAPDH Primer (10 μM)
• Reverse GAPDH Primer (10 μM)
• Nuclease-free water

Store at –20°C.

Format Kit
Reaction Speed 60 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 20 reactions
Reverse Transcriptase RevertAid
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
For Use With (Application) RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 20 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 13 kb
Starting Material RNA
Show More Show Less
When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H-minus RTs for template-independent addition of C nucleotides.

What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.

For Research Use Only. Not for use in diagnostic procedures.