missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pUC57 DNA

Catalog No. FERSD0171
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSD0171 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERSD0171 Supplier Thermo Scientific™ Supplier No. SD0171

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform cloning, sequencing of insert DNA, and generation of nested deletions with ExoIII using this double-stranded closed circular high copy plasmid DNA.

Plasmid pUC57, 2710 bp in length, is a derivative of pUC19. pUC57 MCS contains 6 restriction sites with protruding 3'-ends that are resistant to E. coli exonuclease III. This vector is designed for cloning and generation of ExoIII deletions. The exact positions of the genetic elements are shown on the map (termination codons included). DNA replication initiates at position 890 (±1) and proceeds in the direction indicated. The bla gene nucleotides 2510-2442 (compl. strand) code for a signal peptide.

Highlights

  • More than 90% in the supercoiled form
  • Isolated from E. coli (dam+, dcm+)
  • Purified by chromatography using proprietary patented technology

Specifications

Cloning Method Restriction Enzyme⁄ MCS
Concentration 0.5 μg/μL
Sample Type DNA
Final Product Type DNA
For Use With (Application) Cloning
Product Type DNA
Quantity 50 μg
Vector pUC57

For Research Use Only. Not for use in diagnostic procedures.