missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Genomic DNA Purification Kit

Catalog No. FERK0512
Change view
Click to view available options
Quantity:
100 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0512 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERK0512 Supplier Thermo Scientific™ Supplier No. K0512
  • ONLINE EXCLUSIVE PRICE
    $475.44
    $366.30 / Each

    Save $109.14

    23.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isolate genomic DNA from various sources, including whole blood, serum, cell lines, bacterial cells, plant, and mammalian tissues, with this DNA precipitation system.

Genomic DNA Purification Kit is a simple and rapid system for high quality genomic DNA purification from various sources including: whole blood, serum, cell lines, bacterial cells, plant, and mammalian tissues.

The kit is based on selective detergent-mediated DNA precipitation from crude lysate. The entire procedure is rapid only 20 to 25 minutes with a typical yield of 2 to 10 μg genomic DNA from 0.2 mL of blood.

High molecular weight genomic DNA purified with the kit is suitable for direct use in all common molecular biology applications: PCR, FastDigest and conventional restriction digestion, cloning, DNA sequencing, and Southern blot analysis.

Highlights

  • Efficient—2 to 10 μg of genomic DNA from 0.2 mL of blood
  • Fast—procedure takes approximately 20 minutes
  • Universal—purifies genomic DNA from various sources
  • Flexible—easy to scale up and down
  • Safe—does not involve phenol extraction

Specifications

Content And Storage • 40 mL Lysis Solution; room temperature
• 8 mL Precipitation Solution (concentrated); room temperature
• 10 mL NaCl Solution; room temperature

Isolation Technology Organic Extraction
Sample Type Bacteria, Cells, Plant, Serum, Tissue, Whole Blood, Blood
Elution Volume 20 to 100 μL
Final Product Type gDNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 100 Preps
Shipping Condition Room Temperature
Test Time 25 min.
Starting Material Amount Bacteria: ≤20 mg
Cells: ≤106
Plant: ≤100 mg
Tissue: ≤30 mg
Serum: ≤200 μL
Whole Blood: ≤200 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.