missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Whole Blood Genomic DNA Purification Mini Kit

Catalog No. FERK0782
Change view
Click to view available options
Quantity:
250 Preps
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0782 250 Preps
FERK0781 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0782 Supplier Thermo Scientific™ Supplier No. K0782

Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.

Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.

Highlights
Efficient—high yields of high molecular weight genomic DNA
Fast—20 minute procedure following cell lysis
Convenient—spin columns are capped and assembled with collection tubes
Pure—DNA pure from enzyme inhibitors, RNA and proteins. Ready to use for all demanding applications and validated in real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
Compatible—suitable for wide variety of samples like fresh or frozen anticoagulated mammalian blood (treated with EDTA, Citrate or Heparine), aged blood, buffy coat, bone marrow, and body fluids

Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting Includes Proteinase K Solution Lysis Solution Wash Buffer WB I (concentrated) Wash Buffer II (concentrated) Elution Buffer GeneJET Purification Columns (pre-assembled with collection tubes) Collection tubes Detailed Protocol

Specifications

Column Type Spin Column
Content And Storage • 5 x 1.2 mL Proteinase K Solution; store at room temperature until opened, –20°C after opening
• 120 mL Lysis Solution; room temperature
• 40 mL Wash Buffer I (concentrated); room temperature
• 40 mL Wash Buffer II (concentrated); room temperature
• 150 mL Elution Buffer; room temperature
• 250 GeneJET Purification Columns with Collection Tubes; room temperature
• 250 Collection Tubes; room temperature

Isolation Technology Silica Spin Column
Scale Mini
Sample Type Bone Marrow, Buffy Coat, Dried Blood Spots, Buccal Swab, Urine, Whole Blood
Elution Volume 50 to 200 μL
Final Product Type gDNA
For Use With (Application) PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility Not High-throughput Compatible (Manual)
Quantity 250 Preps
Shipping Condition Room Temperature
Test Time 20 min. (After sample homogenization)
Yield ≤65 μg
Starting Material Amount Bone Marrow: ≤200 μL
Buccal Swabs: 1
Buffy Coat: 200 μL
Dried Blood Spots: varies
Urine: ≤4.5 mL
Whole Blood: ≤500 μL
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Are MagJET and GeneJET blood nucleic acid purification kits compatible with animal blood?

Yes, the kits are compatible with animal blood for DNA/RNA purification.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

For Research Use Only. Not for use in diagnostic procedures.