missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Whole Blood Genomic DNA Purification Mini Kit

Catalog No. FERK0781
Change view
Click to view available options
Quantity:
250 Preps
50 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0781 50 Reactions
FERK0782 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0781 Supplier Thermo Scientific™ Supplier No. K0781

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.

Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.

Highlights
• Efficient—high yields of high molecular weight genomic DNA
• Fast—20 minute procedure following cell lysis
• Convenient—spin columns are capped and assembled with collection tubes
• Pure—DNA pure from enzyme inhibitors, RNA and proteins. Ready to use for all demanding applications and validated in real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
• Compatible—suitable for wide variety of samples like fresh or frozen anticoagulated mammalian blood (treated with EDTA, Citrate or Heparine), aged blood, buffy coat, bone marrow, and body fluids

Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting Includes Proteinase K Solution Lysis Solution Wash Buffer WB I (concentrated) Wash Buffer II (concentrated) Elution Buffer GeneJET Purification Columns (pre-assembled with collection tubes) Collection tubes Detailed Protocol

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Spin Column
Scale Mini
Sample Type Blood
For Use With (Application) Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line GeneJET
Product Type Genomic DNA Purification Kit
Quantity 50 Reactions
Target Genomic DNA
Test Time <20 min.
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Are MagJET and GeneJET blood nucleic acid purification kits compatible with animal blood?

Yes, the kits are compatible with animal blood for DNA/RNA purification.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

For Research Use Only. Not for use in diagnostic procedures.