missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plasmid Miniprep Kit

Catalog No. FERK0502
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0502 50 Preps
FERK0503 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0502 Supplier Thermo Scientific™ Supplier No. K0502

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column.

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.

Highlights

  • Efficient—high yields of up to 20 μg of high quality plasmid DNA
  • Fast—procedure takes less than 14 minutes
  • Convenient—no phenol/chloroform extraction or alcohol precipitation required
  • Pure—purified DNA is immediately ready to use

Applications

Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:

  • FastDigest or conventional restriction digestion
  • Automated fluorescent and radioactive sequencing
  • PCR
  • In vitro transcription
  • Transformation

Principle

A bacterial culture is harvested and lysed. The lysate is then cleared by centrifugation and applied on the silica column to selectively bind DNA molecules at a high salt concentration. The adsorbed DNA is washed to remove contaminants, and the pure plasmid DNA is eluted in a small volume of elution buffer or water. The purified DNA is ready for immediate use in all molecular biology procedures, such as fast and conventional digestion with restriction enzymes, PCR, in vitro transcription, transformation, and automated sequencing.

Components

  • Resuspension Solution
  • Lysis Solution
  • Neutralization Solution Wash Solution (concentrated)
  • RNase A Solution
  • Elution Buffer
  • Spin Columns
  • Collection Tubes
  • Detailed Protocol

Related Products

  • RNase A (Part No.: R1253)
  • Elution Buffer for GeneJET Plasmid Miniprep Kit (Part No.: R1263)
  • Lysis Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1223)
  • Resuspension Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1213)
  • Neutralization Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1233)
  • Wash Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1243)
  • Binding Buffer for GeneJET Plasmid Miniprep Kit(Part No.: R1331)

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Scale Mini
Sample Type Bacteria
For Use With (Application) PCR, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, In Vitro Transcription
High-throughput Compatibility Compatible (Vacuum manifold)
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line GeneJET
Product Type Plasmid MiniPrep Kit
Quantity 50 Preps
Target Plasmid DNA
Test Time <14 min
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Are MagJET and GeneJET plasmid purification kits compatible with EndA+ bacterial strains?

Yes, MagJET and GeneJET Plasmid DNA Purification Kits are suitable for plasmid DNA purification from EndA+ strains. Please refer to the specific kit protocol for details. GeneJET Plasmid Miniprep Kit requires Wash Solution I (Cat. No. R1611, not included in the kit) for EndA+ strains.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

How can I increase plasmid DNA yield during elution from GeneJET plasmid kits?

To increase the overall plasmid DNA yield by 20-30%, additional elution steps may be performed.


For Research Use Only. Not for use in diagnostic procedures.