missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plasmid Midiprep Kit

Catalog No. FERK0482
Change view
Click to view available options
Quantity:
100 Preps
25 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0482 100 Preps
FERK0481 25 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0482 Supplier Thermo Scientific™ Supplier No. K0482

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for large-scale isolation of high quality plasmid DNA from recombinant E.coli cultures.

Thermo Scientific GeneJET Plasmid Midiprep Kit, sufficient for 25 preps, is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. The kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 200 μg of high copy number plasmid DNA. The purified plasmid DNA can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

Highlights

  • Efficient—high yields up to 200 μg of high quality plasmid DNA from 50 mL of bacterial culture
  • High concentration of DNA purified— >400 ng/μL
  • Convenient—spin column format, centrifuge or vacuum protocols
  • Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns

Applications

Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:

  • FastDigest restriction enzyme digestion
  • Automated fluorescent sequencing
  • PCR
  • Cloning
  • Transformation
  • In vitro transcription
  • Transfection of robust cell lines

Specifications

Concentration 400 ng/μL
Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Scale Midi
Sample Type Bacteria
For Use With (Application) Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
Prep Scale 100-200 μg (Medium-Scale) Plasmid DNA
Product Line GeneJET
Product Type Plasmid MidiPrep Kit
Quantity 100 Preps
Target Plasmid DNA
Test Time 60 min.
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Are MagJET and GeneJET plasmid purification kits compatible with EndA+ bacterial strains?

Yes, MagJET and GeneJET Plasmid DNA Purification Kits are suitable for plasmid DNA purification from EndA+ strains. Please refer to the specific kit protocol for details. GeneJET Plasmid Miniprep Kit requires Wash Solution I (Cat. No. R1611, not included in the kit) for EndA+ strains.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

How can I increase plasmid DNA yield during elution from GeneJET plasmid kits?

To increase the overall plasmid DNA yield by 20-30%, additional elution steps may be performed.

Which culture media are recommended for bacteria to be used with GeneJET Plasmid Kits? Can I use a rich growth medium?

The GeneJET Plasmid purification protocols are optimized for bacterial cultures grown in Luria-Bertani (LB) medium. The use of rich growth media is not recommended.

Which bacterial strains can be used for plasmid purification with GeneJET Plasmid KitS?

High quality plasmid DNA can be obtained from various E. coli strains including DH10B, DH5a, XL1-Blue, JM109, JM107, and TOP10.


For Research Use Only. Not for use in diagnostic procedures.