missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plant Genomic DNA Purification Kit

Catalog No. FERK0791
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0791 50 Preps
FERK0792 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0791 Supplier Thermo Scientific™ Supplier No. K0791

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types.

GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Highlights
Fast—procedure completed in 30 min following cell lysis
Convenient—protocol does not include clogging-prone lysate filtration step
High yield—up to 32 μg of high quality DNA from 100 mg sample, including nuclear, mitochondrial, and chloroplast DNA
Compatible—use with awide variety of samples (leaves, roots, sprouts and other parts of various plants)

The standard procedure takes less than 30 minutes following cell lysis and yields purified DNA greater than 30 kb in size. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, and the age of the tissue sample. The typical yield from a standard tissue source (e. g. young wheat leaves) is 30 to 32 μg from 100 mg of tissue. Isolated DNA can be used directly in PCR, qPCR, southern blotting, and enzymatic reactions.

Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Spin Column
Sample Type Plants
For Use With (Application) Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line GeneJET
Product Type Genomic DNA Purification Kit
Quantity 50 Preps
Target Genomic DNA
Test Time <20 min.
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

For Research Use Only. Not for use in diagnostic procedures.