missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET PCR Purification Kit

Catalog No. FERK0702
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0702 250 Preps
FERK0701 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0702 Supplier Thermo Scientific™ Supplier No. K0702
  • ONLINE EXCLUSIVE PRICE
    $623.09
    $465.00 / Each

    Save $158.09

    25.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions. It effectively removes primers, dNTPs, unincorporated labeled nucleotides, enzymes, and salts from PCR and other reaction mixtures.

The kit can be used for purification of DNA fragments from 25 bp to 20 kb with recovery rates up to 100%. Each GeneJET purification column has a total binding capacity of up to 25 μg of DNA, and the entire procedure takes 5 minutes. The purified DNA can be used in common downstream applications, such as sequencing, restriction digestion, labeling, ligation, cloning, in vitro transcription, blotting or in situ hybridization.

Highlights

  • Highly efficient: 90 to 100% recoveries in the range of 100 bp to 10 kb
  • Fast: procedure takes only 5 minutes
  • Convenient: spin columns are capped and assembled with collection tubes

Applications

Fast and efficient purification of DNA fragments ideal for use in all conventional molecular biology procedures including:

  • FastDigest or conventional restriction digestion
  • Automated fluorescent or radioactive sequencing
  • PCR
  • In vitro transcription
  • In situ hybridization
  • Labeling
  • Cloning

Specifications

Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Format Spin Column
Elution Volume Max 50 μL
High-throughput Compatibility Not High-throughput Compatible (Manual)
Purification Time 5 min.
Quantity 250 Preps
Run Time 5 min.
Shipping Condition Room Temperature
Starting Material Volume ≤400 μL
Yield 25 μg (Binding capacity)
For Use With (Application) Nucleic Acid Labeling, Southern Blotting, DNA Sequencing, Nucleic Acid Hybridization, Nucleic Acid Hybridization
Final Product Type DNA
Isolation Technology Silica Spin Column
Sample Type PCR Reactions, Enzymatic Reactions (DNase digestions, RNA labeling and clean-up preps, IVT reactions)
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

Can I use GeneJET PCR Purification Kit for DNA gel extraction?

GeneJET PCR Purification Kit is designed for PCR reaction clean-up. The kit can be used for gel extraction by following the protocol of GeneJET Gel Extraction Kit, but DNA yield may be compromised. For best performance of DNA gel extraction, we recommend to use GeneJET Gel Extraction Kit. For a 3-in-1 kit for purification of PCR products from enzymatic reaction mixtures and agarose gels, we recommend GeneJET Gel Extraction and DNA Cleanup Micro Kit (Cat. Nos. K0831/2).

Can I use GeneJET PCR Purification Kit for enzymatic clean-up?

GeneJET PCR Purification Kit can be used for enzymatic clean-up. Please follow the PCR reaction clean-up protocol.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

Do you offer the binding buffer included in the GeneJET PCR Purification Kit as a stand-alone product?

Yes, we offer Binding Buffer for GeneJET Gel Extraction Kit as a stand-alone product: Cat. No. R1331.


For Research Use Only. Not for use in diagnostic procedures.