missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Gel Extraction and DNA Cleanup Micro Kit

Catalog No. FERK0832
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0832 250 Preps
FERK0831 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0832 Supplier Thermo Scientific™ Supplier No. K0832

The GeneJET Gel Extraction and DNA Cleanup Micro Kit effectively removes primers, primer dimers, dNTPs, unincorporated labeled nucleotides, enzymes and salts from PCR and other reaction mixtures.

The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

The GeneJET Gel Extraction and DNA Cleanup Micro Kit effectively removes primers, primer dimers, dNTPs, unincorporated labeled nucleotides, enzymes and salts from PCR and other reaction mixtures. The kit can be used for purification of DNA fragments from 100 bp to 20 kb. The recovery rates are 90 to 100% for 100 bp to 3 kb DNA fragment size range. Each GeneJET DNA Purification Column has a total binding capacity of up to 10 μg of DNA, and the entire procedure takes approximately 3.5 minutes for DNA cleanup

Thermo Scientific GeneJET Gel Extraction and DNA Cleanup Micro Kit is developed as 3 in 1 kit designed for rapid and efficient purification of DNA from PCR, enzymatic reaction mixtures, and DNA extraction from standard or low-melting point agarose gels run in either Tris acetate (TAE) or Tris borate (TBE) buffer. The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

The GeneJET Gel Extraction and DNA Cleanup Micro Kit effectively removes primers, primer dimers, dNTPs, unincorporated labeled nucleotides, enzymes and salts from PCR and other reaction mixtures. The kit can be used for purification of DNA fragments from 100 bp to 20 kb. The recovery rates are 90 to 100% for 100 bp to 3 kb DNA fragment size range. Each GeneJET DNA Purification Column has a total binding capacity of up to 10 μg of DNA, and the entire procedure takes approximately 3.5 minutes for DNA cleanup

Highlights
Advanced column design—easy to open, load and handle
Yields concentrated DNA - Elute in 6 to 10 μL volume
High recovery - 90 to 100% recovery in the 100 to 3000 bp DNA fragment range
Fast—4 minute reaction mixture clean-up and 15 minute agarose gel purification protocols available
Efficient removal of reaction contaminants - including enzymes, proteins, primers dimers, dNTPs, and salts
Flexible—does not require precise gel slice quantification weighting

Applications
The purified DNA can be used in common downstream applications:
• Sequencing
• Restriction digestion
• PCR
• qPCR
• Labeling
• Ligation
• Cloning
In vitro transcription
• Blotting
In situ hybridization

Includes
• Binding Buffer
• Extraction Buffer
• Prewash Buffer (conc.)
• Wash Buffer (conc.)
• Elution Buffer
• DNA Purification Micro Column & Collection Tube

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Sample Type DNA, Gel Samples
For Use With (Application) Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Nucleic Acid Labeling, Sequencing, PCR, In Vitro Transcription
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line GeneJET
Product Type Gel Extraction and DNA Cleanup Kit
Quantity 250 Preps
Target DNA from Enzymatic Reactions (e.g. PCR)
Test Time 15 min.
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

For Research Use Only. Not for use in diagnostic procedures.