missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq DNA Polymerase (5 U/μL)

Catalog No. FEREP0714
Change view
Click to view available options
Color:
Colorless
Green
Quantity:
10 x 500 Units
20 x 500 Units
200 Units
5 x 500 Units
500 Units
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
FEREP0714 Green 20 x 500 Units
FEREP0701 Colorless 200 Units
FEREP0702 Colorless 500 Units
FEREP0703 Colorless 5 x 500 Units
FEREP0705 Colorless 10 x 500 Units
FEREP0704 Colorless 20 x 500 Units
FEREP0711 Green 200 Units
FEREP0712 Green 500 Units
FEREP0713 Green 5 x 500 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. FEREP0714 Supplier Thermo Scientific™ Supplier No. EP0714

Thermo Scientific DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase.

Thermo Scientific DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase. DreamTaq DNA Polymerase uses the same reaction setup and cycling conditions as conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required. It is supplied with optimized DreamTaq Buffer, which includes 20 mM MgCl2

DreamTaq Green DNA Polymerase is a combination of DreamTaq DNA Polymerase and 10X DreamTaq Green Buffer. The 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with PCR performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications

  • Routine PCR amplification of DNA fragments up to 6 kb
  • High throughput PCR
  • Genotyping

Notes

  • For applications requiring PCR product analysis by absorbance or fluorescence excitation, colorless 10X DreamTaq Buffer (Cat. No. B65) is recommended for purifying PCR products before analysis.

Specifications

Concentration 5 U/μL
Content And Storage • DreamTaq DNA Polymerase (5 U/μL), 20 x 100 μL
• 10X DreamTaq Green Buffer with 20 mM MgCl2, 40 x 1.25 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Product Line DreamTaq
Product Type DreamTaq DNA Polymerase
Quantity 20 x 500 Units
Shipping Condition Dry Ice
For Use With (Application) Standard PCR, DNA Sequencing
Fidelity (vs. Taq) 1X
Hot Start No
Overhang 3'-A
Reaction Format Standalone
Color Green
Show More Show Less
What is the error rate of DreamTaq DNA Polymerases?

The error rate of DreamTaq DNA Polymerase is the same as that of Taq DNA Polymerase: 2.2x10^-5 errors per nt per cycle.

For Research Use Only. Not for use in diagnostic procedures.