missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lambda DNA (dam–, dcm–)

Catalog No. FERSD0021
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSD0021 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERSD0021 Supplier Thermo Scientific™ Supplier No. SD0021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Generate DNA size marker fragments with this linear, double-stranded DNA, a common substrate for restriction endonucleases.

Lambda is a temperate Escherichia coli bacteriophage. The virion DNA is linear and double-stranded (48502 nt) with 12 bp single-stranded complementary 5'-ends. After the phage particle injects its chromosome into the cell, the chromosome circularizes by end joining. In the lytic pathway, phage genes encoding replication, lysis, and virion proteins are expressed. The chromosome replicates, and the replicas are cleaved and packaged into progeny phage particles. In the lysogenic pathway, phage gene expression is repressed, and the circular chromosome inserts into the bacterial chromosome by recombination.

Highlights

Phage lambda DNA is a common substrate for restriction endonucleases and for generating DNA size marker fragments. For large scale isolation of phage DNA, cI857Sam7, a mutant carrying four known mutations, is often used. The DNA sequence used to construct a phage alpha restriction map includes these mutations. The map shows enzymes that cut lambda DNA. Enzymes produced by Thermo Scientific are shown in orange (see Figure bellow).

Lambda DNA (dam-, dcm-) is isolated from a heat-inducible lysogenic E. coli GM2163 strain.

Specifications

Concentration 0.3 μg/μL
Sample Type DNA
Final Product Type DNA
For Use With (Application) CloningNucleic Acid Gel Electrophoresis
Includes [other]
Product Type Lambda DNA
Quantity 500 μg
Species Lambda phage

For Research Use Only. Not for use in diagnostic procedures.