missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CloneJET PCR Cloning Kit

Catalog No. FERK1231
Change view
Click to view available options
No. of Reactions:
20 Reactions
40 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
FERK1231 20 Reactions
FERK1232 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK1231 Supplier Thermo Scientific™ Supplier No. K1231

Efficiently clone PCR products generated with any thermostable DNA polymerase using Thermo Scientific™ CloneJET™ PCR Cloning Kit, a positive selection system.

CloneJET PCR Cloning Kit is an advanced positive selection system for high-efficiency cloning of PCR products generated with any thermostable DNA polymerase. Any other blunt or sticky-end DNA fragment can be cloned. It is ideal for phosphorylated or non-phosphorylated DNA fragments. Ligation into the included positive selection vector takes only 5 minutes, yielding more than 99% recombinant clones. Blunt-ended PCR products generated with a proofreading enzyme are ligated directly into the cloning vector.

PCR products generated either with non-proofreading DNA polymerases or mixtures of DNA polymerases are blunted prior to ligation in 5 minutes with the thermostable DNA Blunting Enzyme provided with the kit. All common laboratory E. coli strains can be directly transformed with the ligation product.

Additionally, any other blunt or sticky-end DNA fragment can be cloned. The kit is ideal for phosphorylated or non-phosphorylated DNA fragments. Ligation into the included positive selection vector takes only 5 minutes and yields more than 99% recombinant clones. Blunt-ended PCR products generated with a proofreading enzyme (e.g., Pfu polymerase) are ligated directly into the cloning vector. PCR products generated either with non-proofreading DNA polymerases (e.g., Taq polymerase) or mixtures of DNA polymerases are blunted prior to ligation in 5 minutes with the thermostable DNA Blunting Enzyme provided with the kit. All common laboratory E.coli strains can be directly transformed with the ligation product.

The kit contains a novel, ready-to-use positive selection cloning vector pJET1.2/blunt. The vector contains a lethal restriction enzyme gene that is disrupted by ligation of a DNA insert into the cloning site. As a result, only bacterial cells with recombinant plasmids are able to form colonies. Recircularized pJET1.2/blunt vector molecules lacking an insert express a lethal restriction enzyme which kills the host E.coli cell after transformation. This positive selection drastically accelerates the process of colony screening and eliminates additional costs required for blue/white selection. For convenience in mapping and manipulation of the insert, the pJET1.2/blunt cloning vector multiple cloning site contains two BglII recognition sequences that flank the insertion site. In addition, the vector contains a T7 promoter for in vitro and in vivo transcription as well as sequencing of the insert.

  • Fast—PCR cloning in only 5 minutes
  • Highest efficiency—>99% of positive clones
  • No cloning background—positive selection vector
  • Versatile—ideal for blunt-end or sticky-end cloning
  • Economical—no expensive blue/white screening

Recommended for:

Cloning of blunt-end or 3'-dA tailed PCR products up to 10kb; Cloning of DNA fragments generated by restriction enzymes; Sequencing of cloned DNA; In vitro and in vivo transcription of cloned inserts from the T7 promoter

Note:

Prior to electroporation always column-purify the ligation mixture using, e.g., GeneJET PCR Purification Kit, †K0701, or chloroform extract kit. Electroporation is inhibited by the presence of proteins and salts in the mixture.

Specifications

Antibiotic Resistance Bacterial Ampicillin (AmpR)
Cloning Method Restriction Enzyme⁄ MCS, Blunt PCR
Format Kit
Sample Type PCR Products
For Use With (Application) PCR Cloning
No. of Reactions 20 Reactions
Product Line CloneJET
Product Type PCR Cloning Kit
Quantity 20 Reactions
Vector pJET1.2/blunt
Can I use the vector pJET1.2 for protein expression?

The pJET1.2 vector is not specifically designed for protein expression. It contains the T7 RNA promoter for in vitro transcription of the cloned insert. In some cases, this T7 promoter can be used for non-toxic gene expression in bacterial strains having IPTG inducible T7 RNA polymerase (like E. coli BL21); however, the promoter does not contain regulatory elements and protein expression will not be well regulated.

What are expected results of colony PCR when screening clones from the CloneJET kit?

Using pJET1.2 Forward and Reverse Sequencing Primers in PCR (as described in the protocol), negative colonies would yield PCR products that are about 120 bp long. Positive colonies would yield longer PCR products that should reflect the insert size (with addition of 120 bp from the vector).

Where can I expect the insert in the pJET1.2/blunt cloning vector after successful cloning?

The insert can be expected at the Eco32I (EcoRV) site in the vector at the position 369 since the pJET1.2/blunt cloning vector was linearized with Eco32I.

What type of DNA fragments can be cloned into the pJET1.2/blunt vector?

Any DNA fragment (either blunt- or sticky-end) that is 6 bp to 10 kb long and generated from PCR or restriction digestion, can be successfully cloned using the kit.

Do I need phosphorylated PCR Primers for CloneJET PCR Cloning Kit?

Phosphorylation of the PCR primers is not required since the 5'-ends of the pJET1.2/blunt vector contain phosphoryl groups for ligation.

For Research Use Only. Not for use in diagnostic procedures.