missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aminoallyl-UTP Solution (50 mM)

Catalog No. FERR1091
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1091 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1091 Supplier Thermo Scientific™ Supplier No. R1091

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Aminoallyl-UTP (5-[3-aminoallyl]-2'-uridine-5'-triphosphate) is supplied as 50 mM aqueous solution titrated to pH 7.0 with NaOH.

Thermo Scientific Aminoallyl-UTP (5-[3-aminoallyl]-2'-uridine-5'-triphosphate) is supplied as 50 mM aqueous solution titrated to pH 7.0 with NaOH.

Aminoallyl-UTP can be enzymatically incorporated into RNA with T7 (Cat. No. EP0111), T3 (EP0101) and SP6 RNA polymerases (EP0131). The resulting amine-containing RNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin, or hapten.

Applications
• Indirect non-radioactive enzymatic labeling of RNA during in vitro transcription

Specifications

Content And Storage Aminoallyl-UTP Solution, 50 mM (50 μL)

Store at –20°C.
Concentration 50 mM
For Use With (Application) Non-Radioactive Labeling
Labeling Method Direct Labeling
Label or Dye Aminoallyl
Product Type Aminoallyl-UTP Solution
Quantity 50 μL
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.