missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TriTrack DNA Loading Dye (6X)
GREENER_CHOICE

Catalog No. ferr1161
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1161 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1161 Supplier Thermo Scientific™ Supplier No. R1161

Prepare DNA markers and samples for loading on agarose or polyacrylamide gels with Thermo Scientific™ 6X TriTrack DNA Loading Dye.

6X TriTrack DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It contains three different dyes (bromophenol blue, xylene cyanol FF or orange G) for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solutions binds divalent metal ions and inhibits metal-dependent nucleases.

6X TriTrack DNA Loading Dye is used for conventional DNA electrophoresis.

Highlights

  • Three-color tracking of DNA migration during DNA electrophoresis 
  • No DNA masking during gel exposure to UV light
  • EDTA binds divalent metal ions and inhibits metal dependent nucleases

Specifications

Concentration 6X
Gel Compatibility Acrylamide Gels, Agarose Gels
Label or Dye Bromophenol Blue, Orange G, Xylene Cyanol FF (XCFF)
Product Type DNA Loading Dye
Quantity 5 x 1 mL

For Research Use Only. Not for use in diagnostic procedures.