missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MgCl2 (magnesium chloride) (25 mM)
Thermo Scientific 25 mM MgCl2 is an aqueous solution of 0.22 μm membrane-filtered magnesium chloride, used for optimization of magnesium ion concentration in PCR.
Supplier: Thermo Scientific™ R0971
Description
Thermo Scientific 25 mM MgCl2 is an aqueous solution of 0.22-μm membrane-filtered magnesium chloride, used for optimization of magnesium ion concentration in PCR with:
- Taq DNA Polymerase (EP0072)
- DreamTaq DNA Polymerase (EP0701)
- Phire Hot Start II DNA Polymerase (F122L).
Features
- Free of endodeoxyribonucleases, exodeoxyribonucleases, ribonucleases, and phosphatases
Applications
- PCR
- Enzymatic reactions
Specifications
| Magnesium Chloride | |
| 25 mM | |
| MgCl2 (Magnesium Chloride) | |
| Wet or Dry Ice |
| • 25 mM MgCl2, 4 x 1.25 mL Store in a non-frost-free freezer at -20°C. |
|
| Hot-start PCR | |
| 4 x 1.25 mL | |
| Liquid |
For Research Use Only. Not for use in diagnostic procedures.