missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenofibric Acid 98.0+%, TCI America™
$52.19 - $167.76
Chemical Identifiers
| CAS | 42017-89-0 |
|---|---|
| Molecular Formula | C17H15ClO4 |
| Molecular Weight (g/mol) | 318.75 |
| MDL Number | MFCD00792461 |
| InChI Key | MQOBSOSZFYZQOK-UHFFFAOYSA-N |
| Synonym | 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid |
| PubChem CID | 64929 |
| ChEBI | CHEBI:83469 |
| IUPAC Name | 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid |
| SMILES | CC(C)(OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1)C(O)=O |
Chemical Identifiers
| 42017-89-0 | |
| 318.75 | |
| MQOBSOSZFYZQOK-UHFFFAOYSA-N | |
| 64929 | |
| 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid |
| C17H15ClO4 | |
| MFCD00792461 | |
| 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid | |
| CHEBI:83469 | |
| CC(C)(OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1)C(O)=O |
Specifications
| 42017-89-0 | |
| White | |
| MFCD00792461 | |
| 3077 | |
| MQOBSOSZFYZQOK-UHFFFAOYSA-N | |
| 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid | |
| 64929 | |
| 318.75 | |
| Crystalline Powder |
| 181°C | |
| C17H15ClO4 | |
| 1 g | |
| 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid | |
| CC(C)(OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1)C(O)=O | |
| 318.75 | |
| CHEBI:83469 | |
| ≥98.0% (GC,T) | |
| Fenofibric Acid |
Safety and Handling
RTECSNumber : UA2453000
TSCA : No