missing translation for 'onlineSavingsMsg'
Learn More

Fendiline hydrochloride

Catalog No. AAJ6325406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6325406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6325406 Supplier Thermo Scientific Chemicals Supplier No. J6325406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13636-18-5 | C23H26ClN | 351.918 g/mol

Fendiline hydrochloride is used as a calcium channel blocker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fendiline hydrochloride is used as a calcium channel blocker.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 13636-18-5
Molecular Formula C23H26ClN
Molecular Weight (g/mol) 351.918
MDL Number MFCD00079301
InChI Key NJXWZWXCHBNOOG-UHFFFAOYSA-N
Synonym fendiline hydrochloride, fendiline hcl, dsstox_cid_25860, dsstox_rid_81180, dsstox_gsid_45860, 3,3-diphenyl-n-1-phenylethyl propan-1-amine hydrochloride, prestwick_568, sensit tn
PubChem CID 5702162
IUPAC Name 3,3-diphenyl-N-(1-phenylethyl)propan-1-amine;hydrochloride
SMILES CC(C1=CC=CC=C1)NCCC(C2=CC=CC=C2)C3=CC=CC=C3.Cl

Specifications

Melting Point 204°C to 205°C
Color White
Quantity 5 g
Merck Index 14,3970
Solubility Information Soluble in ethanol.
Formula Weight 351.92
Physical Form Powder
Chemical Name or Material Fendiline hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 237-121-5
RTECSNumber DP4790000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only