accessibility menu, dialog, popup

UserName

CAS RN 13636-18-5

IUPAC Name: (3,3-diphenylpropyl)(1-phenylethyl)azanium chloride
Synonyms: (3,3-diphenylpropyl)(1-phenylethyl)azanium chloride
Molecular Weight (g/mol): 351.92
Molecular Formula: C23H26ClN
InChi Key: NJXWZWXCHBNOOG-UHFFFAOYNA-N
SMILES: [Cl-].CC([NH2+]CCC(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results