Learn More
Felodipine
A dihydropyridine calcium channel blocker | CAS: 72509-76-3 | C18H19Cl2NO4 | 384.253 g/mol
Supplier: Thermo Scientific Chemicals J61195MD
Description
Felodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFelodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.
Solubility
Soluble in DMSO. Insoluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| Felodipine | |
| Yellow | |
| MFCD00868316 | |
| 14,3953 | |
| Soluble in DMSO; Insoluble in water | |
| CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C | |
| 384.253 | |
| CHEBI:585948 | |
| Crystalline Powder |
| 72509-76-3 | |
| C18H19Cl2NO4 | |
| 250 mg | |
| felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex | |
| RZTAMFZIAATZDJ-UHFFFAOYSA-N | |
| 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate | |
| 3333 | |
| 384.25 |
Chemical Identifiers
| 72509-76-3 | |
| 384.253 | |
| RZTAMFZIAATZDJ-UHFFFAOYSA-N | |
| 3333 | |
| 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| C18H19Cl2NO4 | |
| MFCD00868316 | |
| felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex | |
| CHEBI:585948 | |
| CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : US7968700
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only