missing translation for 'onlineSavingsMsg'
Learn More

Felodipine

Catalog No. p-7042048
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61195MF 50 mg
AAJ61195MC 100 mg
AAJ61195MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61195MF Supplier Thermo Scientific Chemicals Supplier No. J61195MF
Only null left

A dihydropyridine calcium channel blocker | CAS: 72509-76-3 | C18H19Cl2NO4 | 384.253 g/mol

Felodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Felodipine is used as a dihydropyridine calcium channel blocker. It displays high vascular selectivity; lowers arterial blood pressure without altering cardiac contractility. Antihypertensive.

Solubility
Soluble in DMSO. Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 72509-76-3
Molecular Formula C18H19Cl2NO4
Molecular Weight (g/mol) 384.253
MDL Number MFCD00868316
InChI Key RZTAMFZIAATZDJ-UHFFFAOYSA-N
Synonym felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex
PubChem CID 3333
ChEBI CHEBI:585948
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C

Specifications

Color Yellow
Quantity 50 mg
Merck Index 14,3953
Solubility Information Soluble in DMSO; Insoluble in water
Formula Weight 384.25
Physical Form Crystalline Powder
Chemical Name or Material Felodipine
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber US7968700
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only