accessibility menu, dialog, popup

UserName

H3COOH3CHNClClOOH3CCH3

CAS RN 72509-76-3

IUPAC Name: 3-ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: felodipine bc brand of felodipine munobal stadapharm brand of felodipine felo-puren prevex promed brand of felodipine penedil astra brand of felodipine flodil
Molecular Weight (g/mol): 384.25
Molecular Formula: C18H19Cl2NO4
InChi Key: RZTAMFZIAATZDJ-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(Cl)=C1Cl)C(=O)OC
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)