missing translation for 'onlineSavingsMsg'
Learn More

Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™

for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline

$234.45 - $2201.85

Specifications

CAS 1310-73-2,304-59-6,7732-18-5
MDL Number MFCD00012467
UN Number UN1824
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000603
View Documents
Honeywell-Fluka
36019500ML
500 mL
Each for $234.45
Only null left
 
6000599
View Documents
Honeywell-Fluka
360191L
1 L
Each for $420.93
Only null left
 
60046589
View Documents
Honeywell-Fluka
360196X1L
6 x 1 L
Case of 6 Each for $2,201.85
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

1310-73-2,304-59-6,7732-18-5
MFCD00012467
UN1824
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'