missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Fehling’s reagent II for sugars |
| CAS | 1310-73-2, 304-59-6, 7732-18-5 |
| MDL Number | MFCD00012467 |
| UN Number | UN1824 |
| Density | 1.23-1.25g/mL at 20°C |
| Quantity | 1 L |