accessibility menu, dialog, popup

UserName

CAS RN 7758-98-7

IUPAC Name: copper(2+) sulfate
Synonyms: copper sulfate copper(II) sulfate CuSO4 cupric sulfate anhydrous
Molecular Weight (g/mol): 159.6
Molecular Formula: CuO4S
InChi Key: ARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES: [Cu++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (14)
  • (15)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
  • (1)
  • (1)
  • (4)
  • (10)
  • (4)
Physical Form 
  • (2)
  • (3)
  • (1)
  • (8)
  • (7)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (14)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (7)
  • (1)
  • (6)
  • (1)
  • (7)
  • (15)
  • (14)
  • (15)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (10)
  • (4)
  • (2)
  • (3)
  • (1)
  • (8)
  • (7)
  • (1)
  • (1)
  • (1)
  • (3)
  • (14)
  • (4)