missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Red No. 3, Spectrum™ Chemical
FD135, 16423-68-0, C20H6O5I4Na2
$312.63 - $1,936.10
Chemical Identifiers
| CAS | 16423-68-0 |
|---|---|
| Molecular Formula | C20H6I4Na2O5 |
| Molecular Weight (g/mol) | 879.86 |
| MDL Number | MFCD00144257 |
| InChI Key | IINNWAYUJNWZRM-UHFFFAOYSA-L |
| IUPAC Name | disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
| SMILES | [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602626
|
Spectrum Chemical Mfg Cor
FD13525GM |
25 g | Amber Glass Bottle |
N/A
|
|
||||
|
18602627
|
Spectrum Chemical Mfg Cor
FD135100GM |
100 g | Amber Glass Bottle |
N/A
|
|
||||
|
18602628
|
Spectrum Chemical Mfg Cor
FD135500GM |
500 g | Amber Glass Bottle |
N/A
|
|
||||
|
18602629
|
Spectrum Chemical Mfg Cor
FD1355KGBL |
5 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602630
|
Spectrum Chemical Mfg Cor
FD13520KGBL |
20 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical FD and C Red No. 3 is used as a food, drug and cosmetic dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 16423-68-0 | |
| 879.86 | |
| IINNWAYUJNWZRM-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12 |
| C20H6I4Na2O5 | |
| MFCD00144257 | |
| disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
Specifications
| 16423-68-0 | |
| C20H6I4Na2O5 | |
| IINNWAYUJNWZRM-UHFFFAOYSA-L | |
| disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate | |
| Reagent | |
| Maroon | |
| FD and C Red No. 3 |
| 100% | |
| MFCD00144257 | |
| [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12 | |
| 879.86 | |
| Amber Glass Bottle | |
| 25 g |