missing translation for 'onlineSavingsMsg'
Learn More

FD and C Red No. 3, Spectrum™ Chemical

Catalog No. 18602626
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
20 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602626 25 g Amber Glass Bottle
18602627 100 g Amber Glass Bottle
18602628 500 g Amber Glass Bottle
18602629 5 kg Amber Glass Bottle
18602630 20 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602626 Supplier Spectrum Chemical Mfg Cor Supplier No. FD13525GM

FD135, 16423-68-0, C20H6O5I4Na2

Spectrum™ Chemical FD and C Red No. 3 is used as a food, drug and cosmetic dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 16423-68-0
Molecular Formula C20H6I4Na2O5
Molecular Weight (g/mol) 879.86
MDL Number MFCD00144257
InChI Key IINNWAYUJNWZRM-UHFFFAOYSA-L
IUPAC Name disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12

Specifications

CAS Max % 100%
Grade Reagent
Packaging Amber Glass Bottle
Color Maroon
Quantity 25 g
Chemical Name or Material FD and C Red No. 3