Learn More
MP Biomedicals™ FastGro Synthetic, Animal Free, Chemically Defined FBS Replacement
FastGro allows culturing cells in vitro without the use of serum and without any animal or human derived compound.
Supplier: MP Biomedicals™ 092640054
Description
FastGro allows culturing cells in vitro without the use of serum and without any animal or human derived compound. FastGro is a fully defined additive, avoiding the need for serum, for a wide range of cell culture practices. Since FastGro is chemically defined it avoids lot-to-lot variations. All components in FastGro are highly purified and identified chemical compounds.
Culturing cells in a FastGro enriched medium represents a number of benefits as compared to conventional FBS enriched media.
- Due to its chemically defined nature there are no lot-to-lot variations. This skips the need for testing new batches of serum.
- There is no interference of unknown serum compounds in your research.
- Since it doesn't contain hormones nor cytokines it doesn't bias your research data.
- FastGro is save in use, since it doesn't contain animal or human derived materials.
- It can be stored in the refrigerator. No need for thawing before use.
Specifications
| Store at 2 to 8°C | |
| Synthetic sera | |
| 500 mL | |
| Cell Biology | |
| Cell Culture |
| Liquid | |
| FBS Replacement Serum | |
| Sterile | |
| Liver epithelial cells, HaCaT, Primary human hepatocytes, Hybridoma, VERO, HEK293, CHO, HEP G2, HeLa, Human keratinocytes |