missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals FastDNA™ SPIN Kit for Soil

Catalog No. MP116560300
Change view
Click to view available options
No. of Reactions:
100 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116560300 100 Preps
MP116560200 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116560300 Supplier MP Biomedicals Supplier No. 116560300

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FastDNA™ SPIN Kit for Soil is used for rapid isolation of PCR-ready genomic DNA from any environmental sample.

  • The FastDNA™ SPIN Kit for Soil quickly and efficiently isolates bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples.
  • Soil is processed by a FastPrep™ instrument and Lysing Matrix E tubes designed to efficiently lyse all microorganisms.
  • The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications.
  • Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
  • The FastDNA Spin Kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediments, sludge, compost, manure, rhizosphere or waste water.
  • The FastDNA Spin Kit for Soil contains Lysing Matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors.

Specifications

Capacity 2 mL
Concentration Up to 500 mg per prep
Format Kit
Isolation Technology Glass Milk
Sample Type Soil, Feces, Sludge, Wastewater, Environmental, Compost, Manure, Rhizosphere
For Use With (Equipment) FastPrep™ Instrument
For Use With (Application) Isolate bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples, including sediment, sludge, compost, manure, rhizosphere, waste water and more
No. of Reactions 100 Preps
Product Type Nucleic Acid Isolation, Mini-prep
Species Isolate bacterial, fungal, plant, and animal genomic DNA
Target DNA
Test Time 30 min.
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.