missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastCounting™ Cell Counting Kit

For colorimetric quantitation of viable cell number in proliferation and cytotoxicity assays.

$204.17 - $903.21

Specifications

Content And Storage Stored at -20°C for long-term
For Use With (Application) Viability Assay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP092690132
View Documents Promotion Details
MP Biomedicals™
092690132
500 Tests
Each for $903.21
Only null left
 
MP092690131
View Documents Promotion Details
MP Biomedicals™
092690131
100 Tests
Each for $204.17
Only null left
 
Description

Description

The FastCounting cell counting kit provides a sensitive and convenient method to determine cell viability in cell proliferation and cytotoxicity assays. The kit uses a water-soluble tetrazolium salt, WST-8 (i.e. 2-(2-methoxy-4-nitrophenyl)-3-(4-nitrophenyl)-5-(2,4-disulfophenyl)-2H-tetrazolium, monosodium salt) to quantify the number of live cells by producing a yellow color formazan dye upon bio-reduction by dehydrogenase activities in cells. The amount of this formazan dye generated by the bio-reduction inside cells is directly proportional to the number of living cells. The detection sensitivity of FastCounting cell kits is higher than other tetrazolium salts such as MTT, XTT, MTS or WST-1. Due to its stability and extremely low cytotoxicity, the FastCounting cell kit is suitable for longer incubation periods including up to 24 or 48 hours.
Benefits of using FastCounting cell kit:

  • Highest sensitivity dye for cell viability
  • Lowest cytotoxicity among all tetrazolium reagents
  • Simple procedures without thawing reagents
  • Long shelf life and easy storage
Specifications

Specifications

Stored at -20°C for long-term
Viability Assay
Videos
SDS
Documents

Documents