missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastCounting™ Cell Counting Kit

Catalog No. MP092690132
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP092690132 500 Tests
MP092690131 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP092690132 Supplier MP Biomedicals™ Supplier No. 092690132

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For colorimetric quantitation of viable cell number in proliferation and cytotoxicity assays.

The FastCounting cell counting kit provides a sensitive and convenient method to determine cell viability in cell proliferation and cytotoxicity assays. The kit uses a water-soluble tetrazolium salt, WST-8 (i.e. 2-(2-methoxy-4-nitrophenyl)-3-(4-nitrophenyl)-5-(2,4-disulfophenyl)-2H-tetrazolium, monosodium salt) to quantify the number of live cells by producing a yellow color formazan dye upon bio-reduction by dehydrogenase activities in cells. The amount of this formazan dye generated by the bio-reduction inside cells is directly proportional to the number of living cells. The detection sensitivity of FastCounting cell kits is higher than other tetrazolium salts such as MTT, XTT, MTS or WST-1. Due to its stability and extremely low cytotoxicity, the FastCounting cell kit is suitable for longer incubation periods including up to 24 or 48 hours.
Benefits of using FastCounting cell kit:

  • Highest sensitivity dye for cell viability
  • Lowest cytotoxicity among all tetrazolium reagents
  • Simple procedures without thawing reagents
  • Long shelf life and easy storage

Specifications

Content And Storage Stored at -20°C for long-term
For Use With (Application) Viability Assay
Quantity 500 Tests