missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastAP Thermosensitive Alkaline Phosphatase (1 U/μL)

Catalog No. p-4521359
Change view
Click to view available options
Quantity:
1,000 units
300 units
5 x 1,000 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREF0651 1,000 units
FEREF0652 5 x 1,000 units
FEREF0654 300 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FEREF0651 Supplier Thermo Scientific™ Supplier No. EF0651
Only null left

Catalyze the removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins using this thermosensitive alkaline phosphatase .

FastAP Thermosensitive Alkaline Phosphatase catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA, and nucleotides. This enzyme also removes phosphate groups from proteins.

FastAP is a novel alkaline phosphatase, which is active in all Thermo Scientific restriction enzyme buffers as well as in PCR buffers. It dephosphorylates all types of DNA ends (blunt, 5'- and 3'-overhangs) in 10 minutes at 37°C. The enzyme is inactivated in 5 minutes at 75°C . Therefore, removal of alkaline phosphatase is not required prior to ligation.

Highlights

  • Recombinant enzyme
  • Fast dephosphorylation – 10 minutes at 37°C
  • Fast and complete inactivation – 5 minutes at 75°C
  • Simultaneous digestion and dephosphorylation of vector DNA
  • 100% active in restriction enzyme and PCR buffers
  • One protocol for all types of DNA ends:
    • 5’-overhangs
    • 3’-overhangs
    • blunt-ends
    • single nucleotides
  • PCR clean-up in conjunction with Exo I
  • Protein dephosphorylation

Applications

  • Dephosphorylation of cloning vector DNA to prevent recircularization during ligation
  • Simultaneous digestion and dephosphorylation of vector DNA
  • PCR product clean-up: nucleotide degradation prior to sequencing of PCR product
  • Dephosphorylation of nucleic acid 5'-termini prior to labeling with T4 Polynucleotide Kinase
  • Other applications where dephosphorylation of DNA and RNA substrates is necessary
  • Protein dephosphorylation

Note

  • Binding of FastAP Thermosensitive Alkaline Phosphatase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 65°C for 10 minutes, and chill on ice prior to electrophoresis.
  • FastAP Thermosensitive Alkaline Phosphatase is active in all restriction enzyme buffers and may be added directly to digested DNA. Heat inactivation of the restriction enzyme before dephosphorylation reaction is not necessary.

Specifications

Concentration 1 U/μL
Enzyme Phosphatase
Compatible Buffer PCR Buffer, Enzyme Buffer
Quantity 1,000 units
Product Type Alkaline Phosphatase
Do I need to remove the FastAP Thermosensitive Alkaline Phosphatase prior to ligation?

No, removal is not necessary. The enzyme can be inactivated in 5 mins at 75 degrees C.

How long does the dephosphorylation take when using your FastAP Thermosensitive Alkaline Phosphatase?

Dephosphorylation takes place in approximately 10 mins at 37 degrees C.

For Research Use Only. Not for use in diagnostic procedures.