Learn More
Falcon™ Cell Culture Inserts: For Use With 6-Well Plates
GREENER_CHOICE
Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types
$80.63 - $729.93
Specifications
Culture Area 4.2 cm2
Material PET
For Use With (Equipment) 6 Well Plate
Diameter (Metric) Filter 23.1 mm
No. per Pack 6/Pk.
View More Specs
Products 6
Catalog Number Mfr. No. Pore Size Type Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Type Price Quantity & Availability  
08771
GREENER_CHOICE
View Documents Promotion Details
Falcon™
353090
0.4 μm Transparent Membrane
Pack of 6 Each for $80.63
Case of 8 Pack for $645.04
Only null left
0877115
GREENER_CHOICE
View Documents Promotion Details
Falcon™
353493
0.4 μm Translucent Membrane
Case of 48 Each for $729.93
Only null left
0877120
GREENER_CHOICE
View Documents Promotion Details
Falcon™
353093
8 μm Translucent Membrane
Case of 48 Each for $665.81
Only null left
087713
GREENER_CHOICE
View Documents Promotion Details
Falcon™
353091
3 μm Transparent Membrane
Case of 48 Each for $697.68
Only null left
087716
GREENER_CHOICE
View Documents Promotion Details
Falcon™
353092
3 μm Translucent Membrane
Case of 48 Each for $685.34
Only null left
087717
GREENER_CHOICE
View Documents Promotion Details
Falcon™
353102
1 μm Transparent Membrane
Case of 48 Each for $663.41
Only null left
Description
Description
  • Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane
  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12, and 24 well
  • A broad selection of membrane pore sizes, 0.4, 1.0, 3.0, and 8.0 μm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture
Order Info
Individually packaged in color-coded, peel-open medical-style packaging
Specifications
Specifications
4.2 cm2
6 Well Plate
6/Pk.
TC-Treated
Sterile
PET
23.1 mm
48/Cs.
TC-Treated
6