Learn More
Falcon™ Bacteriological Petri Dishes with Lid
GREENER_CHOICE
Manufactured from crystal-grade polystyrene for optical clarity. Falcon™ Bacteriological Petri Dishes are not tissue culture treated for cell attachment for applications where cell attachment is not desired. The surface is hydrophobic.
$505.49 - $721.47
Specifications
Disposable Single-use
Material Polystyrene
Sterility Sterile
Surface Treatment Non-treated
Type Cell Culture Petri Dish
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Culture Area Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Culture Area Price Quantity & Availability  
08757100A
GREENER_CHOICE
Accessories View Documents Promotion Details
Falcon™
351008
35 mm 10 mm 11.78 cm2
Case of 500 Each for $721.47
Only null left
08757100B
GREENER_CHOICE
Accessories View Documents Promotion Details
Falcon™
351007
60 mm 15 mm 21.29 cm2
Case of 500 Each for $564.58
Only null left
08757100D
GREENER_CHOICE
View Documents Promotion Details
Falcon™
351029
100 mm 15 mm 56.5 cm2
Case of 500 Each for $505.49
Only null left
08757148
View Documents Promotion Details
Falcon™
351058
150 mm 15 mm 151 cm2
Case of 100 Each for $616.02
Only null left
Description
Description
These bacteriological Petri dishes are not tissue culture treated for cell attachment; for applications where cell attachment is not desired. Made from crystal-grade polystyrene for optical clarity. Surface is hydrophobic.
  • Durable construction for stable dish manipulation
  • Crystal-grade, virgin polystyrene for optical clarity
  • Sterile
  • Dimensions listed (diameter x height)
Specifications
Specifications
Single-use
Sterile
Cell Culture Petri Dish
Polystyrene
Non-treated