Learn More
Mettler Toledo™ F2 OIML Class Advanced Weight (F2 PL C E)
Single F2 OIML weight includes a calibration certificate and is housed in a plastic box.
$72.16 - $2607.69
Specifications
Product Type Test Weight
Material Stainless Steel
Case Material Plastic
Density 7950 (± 140) kg/m3
Includes With certificate
View More Specs
Products 5
Catalog Number Mfr. No. Weight (Metric) Shape Price Quantity  
Catalog Number Mfr. No. Weight (Metric) Shape Price Quantity  
01912596
View Documents
Mettler Toledo™
30406415
1 mg Sheet
Each for $75.71
Only null left
01912597
View Documents
Mettler Toledo™
30406416
2 mg Sheet
Each for $72.16
Only null left
01912598
View Documents
Mettler Toledo™
30406420
50 mg Sheet
Each for $72.16
Only null left
01912599
View Documents
Mettler Toledo™
30406424
1 g Knob with Adjusting Cavity
Each for $80.15
Only null left
01912600
View Documents
Mettler Toledo™
30406437
20 kg Knob with Adjusting Cavity
Each for $2,607.69
Only null left
Description
Description
  • With calibration certificate
  • Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
  • Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
  • Weight with knob design have an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
Specifications
Specifications
Test Weight
Plastic
With certificate
Stainless Steel
7950 (± 140) kg/m3