missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ezy-Cal™ Count Precision Size Standards

Meet the unique and demanding requirements of the contamination control laboratory in modern industries.

$818.44 - $939.63

Specifications

Concentration 2000 particles/mL ± 10%
Contents Polystyrene microspheres in water
Color White or Colorless
For Use With (Application) Liquid Particle Counter Calibration
Material Polystyrene
View More Specs

Products 9
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
09980226
View Documents Promotion Details
Thermo Scientific™
DUK6002
2 μm
Each for $818.44
Only null left
 
09980227
View Documents Promotion Details
Thermo Scientific™
DUK6005
5 μm
Each for $818.44
Only null left
 
09980228
View Documents Promotion Details
Thermo Scientific™
DUK6010
10.00 ±0.08 μm
Each for $939.63
Only null left
 
09980229
View Documents Promotion Details
Thermo Scientific™
DUK6015
15.02 ±0.15 μm
Each for $884.75
Only null left
 
09980230
View Documents Promotion Details
Thermo Scientific™
6020
20 μm
Each for $871.81
Only null left
 
09980231
View Documents Promotion Details
Thermo Scientific™
6025
25 μm
Each for $871.81
Only null left
 
09980232
View Documents Promotion Details
Thermo Scientific™
6030
30 μm
Each for $871.81
Only null left
 
09980233
View Documents Promotion Details
Thermo Scientific™
DUK6050
50 μm
Each for $818.44
Only null left
 
09980234
View Documents Promotion Details
Thermo Scientific™
DUK6070
70 μm
Each for $871.66
Only null left
 
Description

Description

Meet the unique and demanding requirements of the contamination control laboratory in modern industries with Thermo Scientific™ Ezy-Cal™ Count Precision Size Standards, easy-to-use liquid particle count controls at 2000 particles per mL. Designed for regular use in liquid particle counters to verify instrument calibration before each use, standards assure reproducibility of the particle counter by permitting a continuous record of its performance using a NIST™-traceable microsphere suspension with a precise concentration.

Features:
  • Ready-to-use (no dilution required) particle size standard for validating optical particle counters
  • Magnetic stir bar included in each bottle for clean, convenient and direct sampling
  • Aqueous suspension medium contains a combination of dispersing agents to keep particles from clumping or sticking to flow surfaces in the particle counter
  • Particle diameters traceable to the Standard Meter through the National Institute of Standards and Technology

Recommended for:

Checking the calibration of liquid particle counters.

Specifications

Specifications

2000 particles/mL ± 10%
White or Colorless
Polystyrene
100 mL
Product stable for a minimum of three months
Polystyrene microspheres in water
Liquid Particle Counter Calibration
Count Control Particles
1.59 at 589nm (25°C)
Videos
SDS
Documents

Documents