missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ezetimibe, 98+%, Thermo Scientific Chemicals

Catalog No. AC467692500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467692500 250 mg
AC467690010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467692500 Supplier Thermo Scientific Chemicals Supplier No. 467692500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163222-33-1
Molecular Formula C24H21F2NO3

Specifications

Quantity 250 mg
Formula Weight 409.43
Chemical Name or Material Ezetimibe

RUO – Research Use Only