Learn More
Brady™ EZ Panel Loc Lock Rail
Close panel doors, even when lockouts are in place. The Brady™ EZ Panel Loc System's unique rail system accommodates multiple locks, eliminating the need for hasps during group lockouts.
$14.26 - $75.78
Specifications
Product Type Lock Rail
Color Yellow
Material Fiberglass-reinforced Nylon
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Dimensions (L x W x H) Price Quantity  
Catalog Number Mfr. No. Quantity Dimensions (L x W x H) Price Quantity  
19-103-940
View Documents
Brady™
51256
1 Ea. 4 x 0.8 x 0.7 x in.
Each for $14.26
Only null left
19-103-941
View Documents
Brady™
51258
1 Ea. 8 x 0.8 x 0.7 x in.
Each for $18.46
Only null left
19-103-942
View Documents
Brady™
51260
6 Pk. 8 x 0.8 x 0.7 x in.
Pack of 6 Each for $75.78
Only null left
19-103-943
View Documents
Brady™
51264
2 Pk. 4 x 0.8 x 0.7 x in.
Pack of 2 Each for $33.12
Only null left
Description
Description
  • Unique lock rail system allows panel door to be closed even when lockouts are in place
  • Accommodates multiple locks, eliminating the need for hasps during group lockouts
  • Lock rail design also keeps weight off of the breakers, which protects delicate electrical components from damage
  • Lock rails are made of durable isoplast polymers with acrylic adhesive backing
  • “No shackle”locks with EX Panel Loc keys are sold separately and are especially useful when you want lockout devices to be kept at the point of need
  • A spacing gasket is also available for low profile panels
Specifications
Specifications
Lock Rail
Yellow
Fiberglass-reinforced Nylon
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'