Learn More
Brady™ EZ Panel Loc Lock Rail
Close panel doors, even when lockouts are in place. The Brady™ EZ Panel Loc System's unique rail system accommodates multiple locks, eliminating the need for hasps during group lockouts.
Supplier: Brady™ 51258
Description
- Unique lock rail system allows panel door to be closed even when lockouts are in place
- Accommodates multiple locks, eliminating the need for hasps during group lockouts
- Lock rail design also keeps weight off of the breakers, which protects delicate electrical components from damage
- Lock rails are made of durable isoplast polymers with acrylic adhesive backing
- "No shackle" locks with EX Panel Loc keys are sold separately and are especially useful when you want lockout devices to be kept at the point of need
- A spacing gasket is also available for low profile panels
Specifications
| Lock Rail | |
| Fiberglass-reinforced Nylon | |
| 8 in. |
| Yellow | |
| 8 x 0.8 x 0.7 x in. | |
| 1 Ea. |