Learn More
MilliporeSigma EZ-Fit™ Filtration Unit with Absorbent Pad
Catalog No. EFHAW100M3
Change view
Click to view available options
Grid Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grid Color
EFHAW100M3 Blue
EFHAB100M3 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EFHAW100M3 Supplier MilliporeSigma Supplier No. EFHAW100M3
Only null left
Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing
The EZ-FIT™ filtration unit is a single-use filtration device made of white, gridded, and mixed cellulose esters membrane with a blue base integrating a cellulose support pad.
  • Provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis
  • E-beam irradiated
  • Vented lid covers the sample throughout filtration
  • 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer
Specifications
Disposable Single-use
Material Polystyrene
Sample Volume 100 mL
Sterility Sterile
Grid Color Blue
Type Filtration Unit Without Absorbent Pad