missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma EZ-Fit™ Filtration Unit

Catalog No. EFHAB10BS
Change view
Click to view available options
Grid Color:
Black
White
Packaging:
Bulk
Single
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grid Color Packaging
EFHAB10BS Black Bulk
EFHAW10BS White Bulk
EFHAW10IS White Single
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EFHAB10BS Supplier MilliporeSigma Supplier No. EFHAB10BS

0.45 μm MCE membrane with 100 mL funnel, suitable for solid cultivation

The EZ-FIT™ filtration unit is a single-use filtration device made of gridded, mixed cellulose esters membrane with a pink base. It provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis.

The filtration units are e-beam irradiated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.

Specifications

Disposable Single-use
For Use With (Equipment) EZ-Fit Filtration Manifold
Material Polystyrene
Sample Volume 100 mL
Sterility Sterile
Grid Color Black
Packaging Bulk
Type Filtration Unit