missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma EZ-Fit™ Filtration Unit

0.45 μm MCE membrane with 100 mL funnel, suitable for solid cultivation

$395.00 - $450.00

Specifications

Disposable Single-use
For Use With (Equipment) EZ-Fit Filtration Manifold
Sample Volume 100 mL
Sterility Sterile
Type Filtration Unit
View More Specs

Products 3
Catalog Number Mfr. No. Grid Color Packaging Price Quantity  
Catalog Number Mfr. No. Grid Color Packaging Price Quantity  
EFHAB10BS
View Documents
MilliporeSigma
EFHAB10BS
Black Bulk
Pack of 40 Each for $415.06
Only null left
 
EFHAW10BS
View Documents
MilliporeSigma
EFHAW10BS
White Bulk
Pack of 48 Each for $395.00
Only null left
 
EFHAW10IS
View Documents
MilliporeSigma
EFHAW10IS
White Single
Each for $450.00
Only null left
 
Description

Description

The EZ-FIT™ filtration unit is a single-use filtration device made of gridded, mixed cellulose esters membrane with a pink base. It provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis.

The filtration units are e-beam irradiated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.

Specifications

Specifications

Single-use
100 mL
Filtration Unit
EZ-Fit Filtration Manifold
Sterile
Videos
SDS
Documents

Documents