missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma EZ-Fit™ Filtration Unit

Catalog No. EFGSW25IS
Change view
Click to view available options
Volume (Metric) Working:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Working
EFGSW25IS 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EFGSW25IS Supplier MilliporeSigma Supplier No. EFGSW25IS

0.22 μm MCE membrane with 250 mL funnel, suitable for solid cultivation

The EZ-FIT™ filtration unit is a single-use filtration device made of white, gridded, and mixed cellulose esters membrane with a pink base. It provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis.

The filtration units are e-beam irradiated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.The EZ-FIT™ Azure feature makes the EZ-FIT™ filtration unit even more ergonomic and convenient with transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative funnel assembly design prevents any leaks or sample by-pass of the membrane.

The filtration units conform to international standards and minimize sample residue. The units can be stacked to save the work area. This disposable device is designed to fit onto the EZ-FIT™ Manifold, which features specific EZ-FIT™ Filtration heads.

Specifications

Disposable Single-use
Material Polystyrene
Sterility Sterile
Grid Color White
Type Filtration Unit
Volume (Metric) Working 250 mL
For Use With (Equipment) Microfil Filtration Unit
Sample Volume 250 mL