Learn More
Applied Biosystems™ Extra MagMAX FFPE DNA/RNA Ultra Reagents
Description
This package includes supplemental reagents for use with the MagMAX FFPE DNA/RNA Ultra Kit (Cat. No. A31881). These additional reagents consist of protease digestion buffer, binding solution, and DNA wash buffer and are specifically for use when processing >40 μm sections. See the User Guide for details.
Specifications
Specifications
| Content And Storage | • 10 mL protease digestion buffer, store at room temperature • 38.5 mL binding solution, store at room temperature • 38.5 mL DNA wash buffer, store (at room temperature) |
| Sample Type | FFPE, Fixed Samples |
| For Use With (Application) | Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| Product Line | MagMAX |
| Product Type | FFPE DNA/RNA Ultra Reagent |
| Quantity | 10 mL kit |
| Shipping Condition | Approved for shipment at Room Temperature. |
| Target | Genomic DNA, Total RNA |
Frequently Asked Questions (FAQs)
There is no specific upper limit to curl size for use with the MagMAX FFPE DNA/RNA Ultra kit (Cat. No. A31881). If all of the tissue is able to be submerged in the 200 µL of buffer, then the large volume of protocol should work. Please keep in mind when processing sections that are thicker than 40 µm, additional digestion buffer, binding solution, and wash buffer, may need to be purchased separately (Cat. No. A32796).
For Research Use Only. Not for use in diagnostic procedures.