missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS SYBR™ GreenER™ qPCR Supermix, universal
Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.
$1316.00 - $6000.00
Specifications
| Concentration | 2X |
|---|---|
| Detection Method | SYBR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
11784200
|
Invitrogen™
11784200 |
200 Reactions |
Each for $1,316.00
|
|
|||||
1178401K
|
Invitrogen™
1178401K |
1000 Reactions |
Each for $6,000.00
|
|
|||||
Description
EXPRESS SYBR GreenER qPCR SuperMix Universal comes with separate tubes of ROX reference dye.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
| 2X | |
| Tube | |
| qPCR | |
| Fast or Standard | |
| SYBR GreenER | |
| Gene Expression |
| SYBR | |
| High | |
| Taq DNA Polymerase | |
| 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ | |
| Real Time PCR SYBR Master Mix | |
| DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.