Learn More
Invitrogen™ EXPRESS qPCR Supermix, universal
EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays.
Supplier:  Invitrogen™ 1178501K
Encompass
 View more versions of this product
Catalog No. 1178501K
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Only null left
Description
Description
  • EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for probe-based qPCR assays
  • This kit comes with a separate tube of ROX reference dye
  • EXPRESS qPCR SuperMix kits with premixed ROX reference dye are also available
This kit provides
  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
Chemical Identifiers
Primer-probe
High
Fast or Standard
Gene Expression
Tube
qPCR
Express qPCR Supermix
DNA (Genomic), cDNA
Specifications
EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube.
Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored.
Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Tube
qPCR
Fast or Standard
ROX (Separate Tube)
2,500 reactions
Gene Expression
DNA (Genomic), cDNA
Primer-probe
High
Platinum Taq DNA Polymerase
7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
Express qPCR Supermix
2500 Reactions
2500 Reactions
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.