missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS One-Step SYBR™ GreenER™ Kit, universal
EXPRESS One-Step SYBR GreenER kits are compatible with fast-cycling instruments for your SYBR-based qPCR experiments.
$2290.00 - $9870.00
Specifications
| Content And Storage | EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored. |
|---|---|
| Detection Method | SYBR |
| Format | Tube |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Taq DNA Polymerase |
| Catalog Number | Mfr. No. | Sufficient For | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sufficient For | Price | Quantity | |||||
11780200
|
Invitrogen™
11780200 |
500 Reactions |
Each for $2,290.00
|
|
|||||
1178001K
|
Invitrogen™
1178001K |
2500 Reactions |
Each for $9,870.00
|
|
|||||
Description
This kit comes with a separate tube of ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with premixed ROX reference dye are also available.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed to be stable for 6 months when properly stored.
Specifications
| EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored. |
|
| Tube | |
| Taq DNA Polymerase | |
| 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ | |
| One-Step qRT-PCR Kit |
| SYBR | |
| 1-step RT-qPCR | |
| Fast or Standard | |
| SYBR GreenER | |
| RNA |
For Research Use Only. Not for use in diagnostic procedures.