missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXPRESS One-Step Superscript™ qRT-PCR Kit, with premixed ROX

Next-generation, real-time qRT-PCR reagents designed to complement fast-cycling instruments. Get the turnaround time you want with the data quality you need.

$2,125.00 - $8,700.00

Specifications

Content And Storage EXPRESS One-Step SuperScript™ qRT-PCR SuperMixes are composed of 2 modules: an EXPRESS RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and an EXPRESS qPCR SuperMix module with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed stable for 6 months when properly stored.
Detection Method Primer-probe
Format Tube
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
11791200
Encompass
View Documents Promotion Details
Invitrogen™
11791200
500 Reactions
Each for $2,125.00
Only null left
 
1179101K
Encompass
View Documents Promotion Details
Invitrogen™
1179101K
2500 Reactions
Each for $8,700.00
Only null left
 
Description

Description

EXPRESS One-Step SuperScript™ qRT-PCR SuperMix Kits incorporate proven Invitrogen enzymes now specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for your probe-based qPCR experiments.

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase - complete, rapid activation provides the best sensitivity and reproducibility
  • The One-Step SuperScript™ III Reverse Transcriptase module has a unique formulation to ensure better yield and less PCR inhibition, specifically for one-step qRT-PCR
  • Flexible formats with ROX premixed or separate - designed to work with default instrument protocols on a wide range of high-throughput fast instruments (e.g., AB 7500 Fast, AB 7900 HT Fast, AB StepOne, Roche LightCycler™ 480, Corbett Rotor-Gene™)
  • The only commercially available one-step qRT-PCR kits containing heat-labile UDG - significantly reduced carryover contamination risk with excellent activity from room temperature to approximately 40°C. Due to the thermostability of SuperScript™ III, the cDNA synthesis step is run at 50°C, at which temperature the heat-labile UDG is totally and irreversibly inactivated

Easy to test and switch:
  • EXPRESS kits work with manufacturer's default protocols on fast instruments and come in a convenient and stable SuperMix format for ultimate convenience
  • The kits are offered with either premixed ROX or a separate tube of ROX to fit your instrument needs

Order Info

Guaranteed to be stable for 6 months when properly stored.

Specifications

Specifications

EXPRESS One-Step SuperScript™ qRT-PCR SuperMixes are composed of 2 modules: an EXPRESS RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and an EXPRESS qPCR SuperMix module with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed stable for 6 months when properly stored.
Tube
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
qRT-PCR Kit
RNA
Primer-probe
1-step RT-qPCR
Fast or Standard
SuperScript
Gene Expression
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.